missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals

28.90 € - 96.40 €

Chemical Identifiers

CAS 456-03-1
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000356
InChI Key QIJNVLLXIIPXQT-UHFFFAOYSA-N
Synonym 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
PubChem CID 68004
IUPAC Name 1-(4-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463317
View Documents
Thermo Scientific Alfa Aesar
A16219.09
10 g
28.90 €
10g
Please to purchase this item. Need a web account? Register with us today!
11473317
View Documents
Thermo Scientific Alfa Aesar
A16219.18
50 g
96.40 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

456-03-1
152.168
QIJNVLLXIIPXQT-UHFFFAOYSA-N
68004
CCC(=O)C1=CC=C(C=C1)F
C9H9FO
MFCD00000356
4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
1-(4-fluorophenyl)propan-1-one

Specifications

456-03-1
100°C to 102°C (22 mmHg)
C9H9FO
MFCD00000356
4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
CCC(=O)C1=CC=C(C=C1)F
152.168
152.17
4'-Fluoropropiophenone
1.096
76°C (168°F)
1.5059
1210310
QIJNVLLXIIPXQT-UHFFFAOYSA-N
1-(4-fluorophenyl)propan-1-one
68004
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 207-255-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.