Learn More
4-Fluorocinnamic acid, 99%, Thermo Scientific Chemicals
38.20 € - 109.00 €
Chemical Identifiers
CAS | 459-32-5 |
---|---|
Molecular Formula | C9H7FO2 |
Molecular Weight (g/mol) | 166.15 |
MDL Number | MFCD00004395 |
InChI Key | ISMMYAZSUSYVQG-ZZXKWVIFSA-N |
Synonym | 4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e |
PubChem CID | 1530234 |
IUPAC Name | (E)-3-(4-fluorophenyl)prop-2-enoic acid |
SMILES | C1=CC(=CC=C1C=CC(=O)O)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10646442
|
Thermo Scientific Acros
207110050 |
5 g | Glass bottle |
38.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10656442
|
Thermo Scientific Acros
207110250 |
25 g | Glass bottle |
109.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
459-32-5 | |
166.15 | |
ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
1530234 | |
C1=CC(=CC=C1C=CC(=O)O)F |
C9H7FO2 | |
MFCD00004395 | |
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
(E)-3-(4-fluorophenyl)prop-2-enoic acid |
Specifications
459-32-5 | |
100.0 | |
White | |
99% | |
FC6H4CH=CHCO2H | |
09, II, 395 | |
ISMMYAZSUSYVQG-ZZXKWVIFSA-N | |
(E)-3-(4-fluorophenyl)prop-2-enoic acid | |
1530234 | |
99% | |
4-Fluorocinnamic acid, 99% |
98.5 | |
207.0°C to 210.0°C | |
Authentic | |
C9H7FO2 | |
MFCD00004395 | |
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e | |
C1=CC(=CC=C1C=CC(=O)O)F | |
166.15 | |
166.15 | |
Crystalline Solid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-288-9
- 4-Fluorocinnamic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only