missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobiphenyl, 97+%, Thermo Scientific Chemicals

39.60 € - 127.00 €

Chemical Identifiers

CAS 324-74-3
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.202
MDL Number MFCD00011650
InChI Key RUYZJEIKQYLEGZ-UHFFFAOYSA-N
Synonym 4-fluorobiphenyl, 4-fluoro-1,1'-biphenyl, p-fluorodiphenyl, 4-fluoro-biphenyl, 4-fluorodiphenyl, biphenyl, 4-fluoro, 1,1'-biphenyl, 4-fluoro, para-fluorobiphenyl, 4-fluorophenyl benzene, p-fluorobiphenyl
PubChem CID 9461
IUPAC Name 1-fluoro-4-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307166
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L07170.03
1 g
39.60 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11317166
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L07170.06
5 g
127.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

324-74-3
172.202
RUYZJEIKQYLEGZ-UHFFFAOYSA-N
9461
C1=CC=C(C=C1)C2=CC=C(C=C2)F
C12H9F
MFCD00011650
4-fluorobiphenyl, 4-fluoro-1,1'-biphenyl, p-fluorodiphenyl, 4-fluoro-biphenyl, 4-fluorodiphenyl, biphenyl, 4-fluoro, 1,1'-biphenyl, 4-fluoro, para-fluorobiphenyl, 4-fluorophenyl benzene, p-fluorobiphenyl
1-fluoro-4-phenylbenzene

Specifications

324-74-3
253°C
MFCD00011650
4-fluorobiphenyl, 4-fluoro-1,1'-biphenyl, p-fluorodiphenyl, 4-fluoro-biphenyl, 4-fluorodiphenyl, biphenyl, 4-fluoro, 1,1'-biphenyl, 4-fluoro, para-fluorobiphenyl, 4-fluorophenyl benzene, p-fluorobiphenyl
C1=CC=C(C=C1)C2=CC=C(C=C2)F
172.202
172.2
4-Fluorobiphenyl
72°C to 76°C
C12H9F
2042410
RUYZJEIKQYLEGZ-UHFFFAOYSA-N
1-fluoro-4-phenylbenzene
9461
≥97%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 206-304-1

missing translation for 'rtecsNumber' : DV5291200

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only