missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

Product Code. 15472648
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80143-73-3
Molecular Formula C13H11F2N
Molecular Weight (g/mol) 219.235
MDL Number MFCD03210766
InChI Key OQVIJNAAQBTAAY-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzyl-4-fluorophenyl-amine
PubChem CID 1133337
IUPAC Name 4-fluoro-N-[(4-fluorophenyl)methyl]aniline
SMILES C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F

Specifications

Quantity 250 mg
Formula Weight 219.23
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-(4-fluorobenzyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'