missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Ethylcyclohexene-1-boronic acid, 98%

Product Code. 15429757
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 871329-72-5
Molecular Formula C8H15BO2
MDL Number MFCD07783854
Quantity 250 mg
Synonym 4-ethylcyclohex-1-en-1-yl boronic acid, boronic acid,b-4-ethyl-1-cyclohexen-1-yl, 4-ethylcyclohex-1-en-1-ylboronic acid, acmc-209qgf, 4-ethylcyclohexene-1-boronic acid, 4-ethyl cyclohexen-1-ylboronic acid, 4-ethyl-1-cyclohexenyl boronic acid, 4-ethylcyclohexen-1-yl boronic acid
InChI Key NCXGOXGWYXCMKH-UHFFFAOYSA-N
SMILES B(C1=CCC(CC1)CC)(O)O
IUPAC Name (4-ethylcyclohexen-1-yl)boronic acid
Molecular Weight (g/mol) 154.016
PubChem CID 44886920
Formula Weight 154.02
Percent Purity 98%
Chemical Name or Material 4-Ethylcyclohexene-1-boronic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.