Learn More
4-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals
68.24 € - 182.91 €
Chemical Identifiers
CAS | 5441-51-0 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.20 |
MDL Number | MFCD00001644 |
InChI Key | OKSDJGWHKXFVME-UHFFFAOYSA-N |
Synonym | 4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl |
PubChem CID | 79506 |
IUPAC Name | 4-ethylcyclohexan-1-one |
SMILES | CCC1CCC(=O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337376
|
Thermo Scientific Alfa Aesar
L09363.14 |
25 g |
68.24 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347376
|
Thermo Scientific Alfa Aesar
L09363.22 |
100 g |
182.91 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5441-51-0 | |
126.20 | |
OKSDJGWHKXFVME-UHFFFAOYSA-N | |
79506 | |
CCC1CCC(=O)CC1 |
C8H14O | |
MFCD00001644 | |
4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl | |
4-ethylcyclohexan-1-one |
Specifications
5441-51-0 | |
192°C to 194°C | |
Characteristic | |
1.4515 | |
1924964 | |
OKSDJGWHKXFVME-UHFFFAOYSA-N | |
4-ethylcyclohexan-1-one | |
79506 | |
99% |
0.895 | |
63°C (145°F) | |
C8H14O | |
MFCD00001644 | |
4-ethylcyclohexanone, cyclohexanone, 4-ethyl, 4-ethyl cyclohexanone, 4-ethyl-cyclohexanone, 1-ethyl-4-oxocyclohexane, trans-4-ethylcyclohexanone, acmc-1aru9, ksc273s8l, 5-ethyl-2-oxocyclohexyl | |
CCC1CCC(=O)CC1 | |
126.20 | |
126.2 | |
4-Ethylcyclohexanone |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 226-629-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.