missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycinnamic acid, prediminantly trans, 98+%, Thermo Scientific Chemicals

Product Code. 11302466
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2373-79-7
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00016848
InChI Key DZLOUWYGNATKKZ-VMPITWQZSA-N
Synonym 4-ethoxycinnamic acid, 3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl acrylic acid, 2e-3-4-ethoxyphenyl prop-2-enoic acid, e-3-4-ethoxyphenyl acrylic acid, e-3-4-ethoxyphenyl prop-2-enoic acid, 4-ethoxyphenylacrylic acid, p-ethoxycinnamic acid, ethoxyphenylacrylicacid, pubchem8239
PubChem CID 704218
IUPAC Name (E)-3-(4-ethoxyphenyl)prop-2-enoic acid
SMILES CCOC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 195°C to 199°C
Quantity 25 g
Beilstein 3198068
Formula Weight 192.22
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxycinnamic acid, predominantly trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'