missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

44.50 € - 666.00 €

Chemical Identifiers

CAS 22237-13-4
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD00674028
InChI Key WRQNDLDUNQMTCL-UHFFFAOYSA-N
Synonym 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
PubChem CID 2734351
IUPAC Name (4-ethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC)(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11470088
View Documents
Thermo Scientific Alfa Aesar
B23683.03
1 g
44.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
11480088
View Documents
Thermo Scientific Alfa Aesar
B23683.06
5 g
168.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11490088
View Documents
Thermo Scientific Alfa Aesar
B23683.14
25 g
666.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22237-13-4
165.983
WRQNDLDUNQMTCL-UHFFFAOYSA-N
2734351
B(C1=CC=C(C=C1)OCC)(O)O
C8H11BO3
MFCD00674028
4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
(4-ethoxyphenyl)boronic acid

Specifications

22237-13-4
C8H11BO3
3530408
WRQNDLDUNQMTCL-UHFFFAOYSA-N
(4-ethoxyphenyl)boronic acid
2734351
98%
165°C to 168°C
MFCD00674028
4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
B(C1=CC=C(C=C1)OCC)(O)O
165.983
165.99
4-Ethoxybenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.