Learn More
4'-Ethoxyacetophenone, 98%, Thermo Scientific Chemicals
27.90 € - 242.00 €
Chemical Identifiers
CAS | 1676-63-7 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.20 |
MDL Number | MFCD00009095 |
InChI Key | YJFNFQHMQJCPRG-UHFFFAOYSA-N |
Synonym | 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone |
PubChem CID | 72872 |
IUPAC Name | 1-(4-ethoxyphenyl)ethanone |
SMILES | CCOC1=CC=C(C=C1)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468863
|
Thermo Scientific Alfa Aesar
A11031.06 |
5 g |
27.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478863
|
Thermo Scientific Alfa Aesar
A11031.14 |
25 g |
75.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11488863
|
Thermo Scientific Alfa Aesar
A11031.22 |
100 g |
242.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.
Solubility
Soluble in alcohol, water(791.1 mg/L).
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
1676-63-7 | |
164.20 | |
YJFNFQHMQJCPRG-UHFFFAOYSA-N | |
72872 | |
CCOC1=CC=C(C=C1)C(C)=O |
C10H12O2 | |
MFCD00009095 | |
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone | |
1-(4-ethoxyphenyl)ethanone |
Specifications
1676-63-7 | |
145°C to 146°C (13 mmHg) | |
C10H12O2 | |
636783 | |
Soluble in alcohol,water(791.1mg/L). | |
CCOC1=CC=C(C=C1)C(C)=O | |
164.20 | |
164.2 | |
4'-Ethoxyacetophenone |
35°C to 38°C | |
134°C (273°F) | |
MFCD00009095 | |
4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone | |
YJFNFQHMQJCPRG-UHFFFAOYSA-N | |
1-(4-ethoxyphenyl)ethanone | |
72872 | |
98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 216-825-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only