missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15459677
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201802-67-7
Molecular Formula C18H16BNO2
Molecular Weight (g/mol) 289.141
MDL Number MFCD06798117
InChI Key TWWQCBRELPOMER-UHFFFAOYSA-N
Synonym 4-diphenylamino phenylboronic acid, triphenylamine-4-boronic acid, 4-diphenylamino phenyl boronic acid, 4-diphenylamino benzeneboronic acid, 4-n,n-diphenylamino phenylboronic acid, boronic acid, b-4-diphenylamino phenyl, 4-n-diphenylamino phenylboronic acid, 4-diphenylamino phenylboronicacid, boronic acid, 4-diphenylamino phenyl
PubChem CID 12166934
IUPAC Name [4-(N-phenylanilino)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 110°C to 115°C
Quantity 1 g
Formula Weight 289.14
Percent Purity 98%
Chemical Name or Material 4-(Diphenylamino)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.