missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

37.29 € - 478.38 €

Chemical Identifiers

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.34
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15432289
View Documents
Thermo Scientific Alfa Aesar
H64887.03
1 g
37.29 €
1g
Please to purchase this item. Need a web account? Register with us today!
15442289
View Documents
Thermo Scientific Alfa Aesar
H64887.06
5 g
126.63 €
5g
Please to purchase this item. Need a web account? Register with us today!
15452289
View Documents
Thermo Scientific Alfa Aesar
H64887.14
25 g
478.38 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4181-05-9
273.34
UESSERYYFWCTBU-UHFFFAOYSA-N
77846
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
C19H15NO
MFCD00145131
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
4-(N-phenylanilino)benzaldehyde

Specifications

4181-05-9
Yellow
C19H15NO
2732795
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
UESSERYYFWCTBU-UHFFFAOYSA-N
4-(N-phenylanilino)benzaldehyde
77846
98%
4-(Diphenylamino)benzaldehyde
129°C to 133°C (literature)
Odorless
MFCD00145131
Air sensitive
Soluble in methanol.
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
273.34
273.33
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.