missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

75.10 € - 298.00 €

Chemical Identifiers

CAS 2131-64-8
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00041093
InChI Key HRDJPEMAGYHSJR-UHFFFAOYSA-N
PubChem CID 75047
IUPAC Name 4-isothiocyanato-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=C=S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312836
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L09011.04
2 g
75.10 €
2g
Please to purchase this item. Need a web account? Register with us today!
11322836
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L09011.09
10 g
298.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2131-64-8
178.253
HRDJPEMAGYHSJR-UHFFFAOYSA-N
4-isothiocyanato-N,N-dimethylaniline
C9H10N2S
MFCD00041093
75047
CN(C)C1=CC=C(C=C1)N=C=S

Specifications

2131-64-8
152°C (6 mmHg)
C9H10N2S
UN2928
Moisture sensitive
HRDJPEMAGYHSJR-UHFFFAOYSA-N
4-isothiocyanato-N,N-dimethylaniline
75047
97%
64°C to 70°C
Irritating
MFCD00041093
388270
4-dimethylamino phenyl isothiocyanate, 4-dimethylaminophenyl isothiocyanate, 4-n,n-dimethylamino phenyl isothiocyanate, p-dimethylamino phenyl isothiocyanate, benzenamine, 4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylaminophenyl ester, p-n-dimethylamino phenyl isothiocyanate, 4-dimethylamino phenylisothiocyanate, benzenamine,4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylamino phenyl ester
CN(C)C1=CC=C(C=C1)N=C=S
178.253
178.25
4-Dimethylaminophenyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 218-360-4

missing translation for 'rtecsNumber' : NX8500000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Certificates

RUO – Research Use Only