Learn More
4-(Dimethylamino)phenethyl alcohol, 99%, Thermo Scientific Chemicals
70.30 € - 243.00 €
Chemical Identifiers
CAS | 50438-75-0 |
---|---|
InChI Key | CDTPAAZQBPSVGS-UHFFFAOYSA-N |
Synonym | 2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol |
PubChem CID | 93252 |
IUPAC Name | 2-[4-(dimethylamino)phenyl]ethanol |
SMILES | CN(C)C1=CC=C(C=C1)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11310800
|
Thermo Scientific Acros
210930010 |
1 g | Glass bottle |
70.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995271
|
Thermo Scientific Acros
210930050 |
5 g | Glass bottle |
243.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
50438-75-0 | |
2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol | |
2-[4-(dimethylamino)phenyl]ethanol |
CDTPAAZQBPSVGS-UHFFFAOYSA-N | |
93252 | |
CN(C)C1=CC=C(C=C1)CCO |
Specifications
50438-75-0 | |
100.0 | |
White to Brown | |
Authentic | |
C10H15NO | |
2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol | |
CN(C)C1=CC=C(C=C1)CCO | |
165.23 | |
165.23 | |
Crystalline Powder |
98.5 | |
53.9°C to 55.1°C | |
116.0°C (0.1 mmHg) | |
98.5% min. (GC) | |
(CH3)2NC6H4CH2CH2OH | |
CDTPAAZQBPSVGS-UHFFFAOYSA-N | |
2-[4-(dimethylamino)phenyl]ethanol | |
93252 | |
99% | |
4-(Dimethylamino)phenethyl alcohol |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 4-(Dimethylamino)phenethyl alcohol, 99%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only