missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzonitrile, 95%, Thermo Scientific Chemicals

Product Code. 10756141
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001815
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 70.0°C to 76.0°C
Color Brown
Boiling Point 318.0°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CN
Quantity 25 g
Beilstein 14, 428
Formula Weight 146.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)benzonitrile, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'