Learn More
4-(Cyclopropylamino)benzonitrile, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 1019607-55-6 |
---|---|
Molecular Formula | C10H10N2 |
Molecular Weight (g/mol) | 158.204 |
InChI Key | SHZXUUVOVQTNCK-UHFFFAOYSA-N |
Synonym | 4-cyclopropylamino benzonitrile, acmc-2097xs |
PubChem CID | 28411077 |
IUPAC Name | 4-(cyclopropylamino)benzonitrile |
SMILES | C1CC1NC2=CC=C(C=C2)C#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1019607-55-6 | |
158.204 | |
4-cyclopropylamino benzonitrile, acmc-2097xs | |
4-(cyclopropylamino)benzonitrile |
C10H10N2 | |
SHZXUUVOVQTNCK-UHFFFAOYSA-N | |
28411077 | |
C1CC1NC2=CC=C(C=C2)C#N |
Specifications
1019607-55-6 | |
UN3439 | |
SHZXUUVOVQTNCK-UHFFFAOYSA-N | |
4-(cyclopropylamino)benzonitrile | |
28411077 | |
98% | |
4-(Cyclopropylamino)benzonitrile |
C10H10N2 | |
4-cyclopropylamino benzonitrile, acmc-2097xs | |
C1CC1NC2=CC=C(C=C2)C#N | |
158.204 | |
158.2 | |
Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures