Learn More
4-Cyanobenzeneboronic acid, 98%, Thermo Scientific Chemicals
104.52 € - 350.75 €
Chemical Identifiers
CAS | 126747-14-6 |
---|---|
Molecular Formula | C7H6BNO2 |
Molecular Weight (g/mol) | 146.94 |
MDL Number | MFCD01318968 |
InChI Key | CEBAHYWORUOILU-UHFFFAOYSA-N |
Synonym | 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid |
PubChem CID | 2734326 |
IUPAC Name | (4-cyanophenyl)boronic acid |
SMILES | OB(O)C1=CC=C(C=C1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374865
|
Thermo Scientific Alfa Aesar
L18007.03 |
1 g |
104.52 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384865
|
Thermo Scientific Alfa Aesar
L18007.06 |
5 g |
350.75 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
126747-14-6 | |
146.94 | |
CEBAHYWORUOILU-UHFFFAOYSA-N | |
2734326 | |
OB(O)C1=CC=C(C=C1)C#N |
C7H6BNO2 | |
MFCD01318968 | |
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid | |
(4-cyanophenyl)boronic acid |
Specifications
126747-14-6 | |
C7H6BNO2 | |
6593772 | |
CEBAHYWORUOILU-UHFFFAOYSA-N | |
(4-cyanophenyl)boronic acid | |
2734326 | |
98% |
>300°C | |
MFCD01318968 | |
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid | |
OB(O)C1=CC=C(C=C1)C#N | |
146.94 | |
146.94 | |
4-Cyanobenzeneboronic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only