Learn More
4-chlorotoluene, 98%, Thermo Scientific Chemicals
19.76 € - 363.81 €
Chemical Identifiers
CAS | 106-43-4 |
---|---|
Molecular Formula | C7H7Cl |
Molecular Weight (g/mol) | 126.58 |
MDL Number | MFCD00000631 |
InChI Key | NPDACUSDTOMAMK-UHFFFAOYSA-N |
Synonym | 4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n |
PubChem CID | 7810 |
ChEBI | CHEBI:34401 |
IUPAC Name | 1-chloro-4-methylbenzene |
SMILES | CC1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10562161
|
Thermo Scientific Acros
150210050 |
5 mL | Glass bottle | N/A | |||||
10229313
|
Thermo Scientific Acros
150210250 |
25 mL | Glass bottle |
25.53 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10086003
|
Thermo Scientific Acros
150211000 |
100 mL | Glass bottle |
60.60 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306460
|
Thermo Scientific Acros
150210010 |
1 L | Glass bottle |
363.81 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-43-4 | |
126.58 | |
NPDACUSDTOMAMK-UHFFFAOYSA-N | |
7810 | |
1-chloro-4-methylbenzene |
C7H7Cl | |
MFCD00000631 | |
4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n | |
CHEBI:34401 | |
CC1=CC=C(Cl)C=C1 |
Specifications
6.0°C to 8.0°C | |
97.5 | |
1.0700g/mL | |
Authentic | |
1.5190 to 1.5210 | |
1.07 | |
CH3C6H4Cl | |
05, 292 | |
4-chlorotoluene, p-chlorotoluene, benzene, 1-chloro-4-methyl, p-tolyl chloride, toluene, p-chloro, 1-methyl-4-chlorobenzene, 4-chloro-1-methylbenzene, monochlorotoluene, para-chlorotoluene, unii-q8r236h42n | |
NPDACUSDTOMAMK-UHFFFAOYSA-N | |
1-chloro-4-methylbenzene | |
7810 | |
126.59 | |
4-Chlorotoluene |
106-43-4 | |
100.0 | |
162.0°C | |
Liquid | |
97.5% min. (GC) | |
C7H7Cl | |
MFCD00000631 | |
15, 2175 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,chloroform and ether | |
CC1=CC=C(Cl)C=C1 | |
126.58 | |
CHEBI:34401 | |
98% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-397-
RUO – Research Use Only