Learn More
4-Chloroquinoline, 99%, Thermo Scientific Chemicals
99.90 € - 379.00 €
Chemical Identifiers
CAS | 611-35-8 |
---|---|
Molecular Formula | C9H6ClN |
Molecular Weight (g/mol) | 163.60 |
MDL Number | MFCD00006773 |
InChI Key | KNDOFJFSHZCKGT-UHFFFAOYSA-N |
Synonym | quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa |
PubChem CID | 69140 |
IUPAC Name | 4-chloroquinoline |
SMILES | ClC1=C2C=CC=CC2=NC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453248
|
Thermo Scientific Alfa Aesar
H56721.03 |
1 g |
99.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15463248
|
Thermo Scientific Alfa Aesar
H56721.06 |
5 g |
379.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Partly soluble in water.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Chemical Identifiers
611-35-8 | |
163.60 | |
KNDOFJFSHZCKGT-UHFFFAOYSA-N | |
69140 | |
ClC1=C2C=CC=CC2=NC=C1 |
C9H6ClN | |
MFCD00006773 | |
quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa | |
4-chloroquinoline |
Specifications
611-35-8 | |
1.25 | |
113°C (235°F) | |
MFCD00006773 | |
quinoline, 4-chloro, 4-chloro-quinoline, ccris 3979, 4-chioroquinoline, acmc-209mpi, 4-chloroquinoline, ksc354e5n, 4-chloroquinoline 1g, kndofjfshzckgt-uhfffaoysa | |
KNDOFJFSHZCKGT-UHFFFAOYSA-N | |
4-chloroquinoline | |
69140 | |
99% |
28°C to 31°C | |
260°C to 261°C | |
C9H6ClN | |
114721 | |
Partly soluble in water. | |
ClC1=C2C=CC=CC2=NC=C1 | |
163.60 | |
163.6 | |
4-Chloroquinoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-267-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.