missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(2-fluorobenzyl)aniline, 97%, Thermo Scientific™

Product Code. 15472728
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723753-84-2
Molecular Formula C13H11ClFN
Molecular Weight (g/mol) 235.69
MDL Number MFCD05740323
InChI Key QCLJXZCWBRKJHD-UHFFFAOYSA-N
Synonym 4-chloro-n-2-fluorophenyl methyl aniline, n-4-chlorophenyl-n-2-fluorobenzyl amine, 4-chloro-n-2-fluorobenzyl aniline
PubChem CID 834588
IUPAC Name 4-chloro-N-[(2-fluorophenyl)methyl]aniline
SMILES FC1=CC=CC=C1CNC1=CC=C(Cl)C=C1

Specifications

Quantity 1 g
Formula Weight 235.69
Percent Purity 97%
Chemical Name or Material 4-Chloro-N-(2-fluorobenzyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'