missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-8-fluoroquinoline, Thermo Scientific Chemicals

130.00 € - 575.00 €

Chemical Identifiers

CAS 63010-72-0
Molecular Formula C9H5ClFN
Molecular Weight (g/mol) 181.59
MDL Number MFCD00278785
InChI Key NUIMKHDPMIPYFI-UHFFFAOYSA-N
PubChem CID 2736587
IUPAC Name 4-chloro-8-fluoroquinoline
SMILES FC1=C2N=CC=C(Cl)C2=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379999
View Documents
Thermo Scientific Alfa Aesar
H50541.03
1 g
130.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11389999
View Documents
Thermo Scientific Alfa Aesar
H50541.06
5 g
575.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

4-Chloro-8-fluoroquinoline is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-8-fluoroquinoline is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

63010-72-0
181.59
NUIMKHDPMIPYFI-UHFFFAOYSA-N
4-chloro-8-fluoroquinoline
C9H5ClFN
MFCD00278785
2736587
FC1=C2N=CC=C(Cl)C2=CC=C1

Specifications

63010-72-0
MFCD00278785
NUIMKHDPMIPYFI-UHFFFAOYSA-N
4-chloro-8-fluoroquinoline
2736587
4-Chloro-8-fluoroquinoline
C9H5ClFN
4-chloro-8-fluoro-quinoline, 8-fluoro-4-chloroquinoline, quinoline, 4-chloro-8-fluoro, pubchem5886, 4-chloro-8fluoroquinoline, 4-chloro-8-fluoroquinolline, quinoline,4-chloro-8-fluoro, 4-chloranyl-8-fluoranyl-quinoline
FC1=C2N=CC=C(Cl)C2=CC=C1
181.59
181.6
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.