missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-4'-fluorobutyrophenone, 96%, Thermo Scientific Chemicals

Product Code. 11439013
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3874-54-2
Molecular Formula C10H10ClFO
Molecular Weight (g/mol) 200.637
MDL Number MFCD00001007
InChI Key HXAOUYGZEOZTJO-UHFFFAOYSA-N
Synonym 4-chloro-4'-fluorobutyrophenone, 4-chloro-1-4-fluorophenyl butan-1-one, 3-4-fluorobenzoyl propyl chloride, 4-fluorobenzoylpropyl chloride, 4-chloro-p-fluorobutyrophenone, 3-chloropropyl 4-fluorophenyl ketone, 1-butanone, 4-chloro-1-4-fluorophenyl, 4'-fluoro-4-chlorobutyrophenone, p-fluorobenzoylpropyl chloride, p-fluoro-4-chlorobutyrophenone
PubChem CID 19750
IUPAC Name 4-chloro-1-(4-fluorophenyl)butan-1-one
SMILES C1=CC(=CC=C1C(=O)CCCCl)F

Specifications

Melting Point 5°C to 6°C
Density 1.23
Boiling Point 120°C to 122°C (0.7 mmHg)
Flash Point 152°C (306°F)
Refractive Index 1.5255
Quantity 100 g
Beilstein 608741
Formula Weight 200.64
Percent Purity 96%
Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'