missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-nitrophenol, 98%, contains up to ca 10% water, Thermo Scientific Chemicals

Product Code. 11433667
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-64-5
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00007113
InChI Key NWSIFTLPLKCTSX-UHFFFAOYSA-N
Synonym 2-nitro-4-chlorophenol, phenol, 4-chloro-2-nitro, unii-438lq62wnh, 4-chloro-2-nitro-phenol, 4-chloro-6-nitrophenol, acmc-209r2b, 5-chloro-2-hydroxynitrobenzene, 4-chloro-2-nitrophenol
PubChem CID 6980
IUPAC Name 4-chloro-2-nitrophenol
SMILES OC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 85°C to 88°C
Quantity 500 g
Beilstein 2048031
Formula Weight 173.56
Percent Purity 98%
Assay contains up to ≈10% water
Chemical Name or Material 4-Chloro-2-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'