Learn More
4-chloro-2-nitroaniline, 99%, Thermo Scientific Chemicals
32.30 € - 225.12 €
Chemical Identifiers
CAS | 89-63-4 |
---|---|
Molecular Formula | C6H5ClN2O2 |
Molecular Weight (g/mol) | 172.57 |
MDL Number | MFCD00007836 |
InChI Key | PBGKNXWGYQPUJK-UHFFFAOYSA-N |
Synonym | benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi |
PubChem CID | 6979 |
IUPAC Name | 4-chloro-2-nitroaniline |
SMILES | NC1=CC=C(Cl)C=C1[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10167753
|
Thermo Scientific Acros
146521000 |
100 g | Glass bottle | N/A | |||||
10615772
|
Thermo Scientific Acros
146520010 |
1 kg | Glass bottle |
225.12 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89-63-4 | |
172.57 | |
PBGKNXWGYQPUJK-UHFFFAOYSA-N | |
6979 | |
NC1=CC=C(Cl)C=C1[N+]([O-])=O |
C6H5ClN2O2 | |
MFCD00007836 | |
benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi | |
4-chloro-2-nitroaniline |
Specifications
89-63-4 | |
100.0 | |
Orange | |
Authentic | |
C6H5ClN2O2 | |
MFCD00007836 | |
benzenamine, 4-chloro-2-nitro, p-chloro-o-nitroaniline, pcon, 2-nitro-4-chloroaniline, red salt nbgl, devol red f, devol red salt f, red base ciba vi, red base irga vi, red salt ciba vi | |
PBGKNXWGYQPUJK-UHFFFAOYSA-N | |
4-chloro-2-nitroaniline | |
6979 | |
99% | |
4-chloro-2-nitroaniline |
98.5 | |
115°C to 119°C | |
191°C | |
98.5% min. (GC) | |
ClC6H3(NO2)NH2 | |
12, 729 | |
Solubility in water: insoluble in water. Other solubilities: soluble in methanol and ether | |
NC1=CC=C(Cl)C=C1[N+]([O-])=O | |
172.57 | |
172.57 | |
Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Fatal if swallowed.
Fatal if inhaled.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-925-4
- 4-Chloro-2-nitroaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only