Learn More
4-Chloro-2-methylphenol, 97%, Thermo Scientific Chemicals
51.05 € - 179.90 €
Chemical Identifiers
CAS | 1570-64-5 |
---|---|
Molecular Formula | C7H7ClO |
Molecular Weight (g/mol) | 142.582 |
MDL Number | MFCD00002321 |
InChI Key | RHPUJHQBPORFGV-UHFFFAOYSA-N |
Synonym | 4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri |
PubChem CID | 14855 |
ChEBI | CHEBI:1800 |
IUPAC Name | 4-chloro-2-methylphenol |
SMILES | CC1=C(C=CC(=C1)Cl)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316826
|
Thermo Scientific Alfa Aesar
L04164.06 |
5 g |
51.05 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326826
|
Thermo Scientific Alfa Aesar
L04164.14 |
25 g |
179.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1570-64-5 | |
142.582 | |
RHPUJHQBPORFGV-UHFFFAOYSA-N | |
14855 | |
4-chloro-2-methylphenol |
C7H7ClO | |
MFCD00002321 | |
4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri | |
CHEBI:1800 | |
CC1=C(C=CC(=C1)Cl)O |
Specifications
1570-64-5 | |
1.2 | |
78°C (172°F) | |
MFCD00002321 | |
1906684 | |
RHPUJHQBPORFGV-UHFFFAOYSA-N | |
4-chloro-2-methylphenol | |
14855 | |
142.59 | |
4-Chloro-2-methylphenol |
45°C to 48°C | |
220°C to 225°C | |
C7H7ClO | |
UN3437 | |
4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri | |
CC1=C(C=CC(=C1)Cl)O | |
142.582 | |
CHEBI:1800 | |
97% |
Safety and Handling
GHS H Statement
H331-H314
Toxic if inhaled.
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H331
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
missing translation for 'einecsNumber' : 216-381-3
missing translation for 'rtecsNumber' : GO7120000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.