missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

84.00 € - 273.00 €

Chemical Identifiers

CAS 762287-57-0
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.398
MDL Number MFCD02684317
InChI Key NZRRMTBNTSBIFH-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxyphenyl boronic acid, 4-chloro-2-methoxybenzeneboronic acid, 4-chloro-2-methoxy-phenyl boronic acid, boronic acid, 4-chloro-2-methoxyphenyl, acmc-209p2g, ksc495a8j, ablock ab-12-6332, 2-methoxy-4-chlorophenylboronic acid, 4-chloranyl-2-methoxy-phenyl boronic acid, 4-chloro-2-methyloxy phenyl boronic acid
PubChem CID 3858606
IUPAC Name (4-chloro-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)OC)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15449267
View Documents
Thermo Scientific Alfa Aesar
H52529.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15439267
View Documents
Thermo Scientific Alfa Aesar
H52529.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

762287-57-0
186.398
NZRRMTBNTSBIFH-UHFFFAOYSA-N
3858606
B(C1=C(C=C(C=C1)Cl)OC)(O)O
C7H8BClO3
MFCD02684317
4-chloro-2-methoxyphenyl boronic acid, 4-chloro-2-methoxybenzeneboronic acid, 4-chloro-2-methoxy-phenyl boronic acid, boronic acid, 4-chloro-2-methoxyphenyl, acmc-209p2g, ksc495a8j, ablock ab-12-6332, 2-methoxy-4-chlorophenylboronic acid, 4-chloranyl-2-methoxy-phenyl boronic acid, 4-chloro-2-methyloxy phenyl boronic acid
(4-chloro-2-methoxyphenyl)boronic acid

Specifications

762287-57-0
C7H8BClO3
4-chloro-2-methoxyphenyl boronic acid, 4-chloro-2-methoxybenzeneboronic acid, 4-chloro-2-methoxy-phenyl boronic acid, boronic acid, 4-chloro-2-methoxyphenyl, acmc-209p2g, ksc495a8j, ablock ab-12-6332, 2-methoxy-4-chlorophenylboronic acid, 4-chloranyl-2-methoxy-phenyl boronic acid, 4-chloro-2-methyloxy phenyl boronic acid
B(C1=C(C=C(C=C1)Cl)OC)(O)O
186.398
186.4
4-Chloro-2-methoxybenzeneboronic acid
128°C to 134°C
MFCD02684317
NZRRMTBNTSBIFH-UHFFFAOYSA-N
(4-chloro-2-methoxyphenyl)boronic acid
3858606
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.