Learn More
4-Bromoisoxazole, 95%, Thermo Scientific Chemicals
29.45 € - 91.10 €
Chemical Identifiers
CAS | 97925-43-4 |
---|---|
Molecular Formula | C3H2BrNO |
Molecular Weight (g/mol) | 147.96 |
InChI Key | IRDVIGFQMLUJAM-UHFFFAOYSA-N |
Synonym | 4-bromoisoxazole, 4-bormoisoxazole, isoxazole, 4-bromo, isoxazole, 4-bromo-6ci,9ci, 4-bromo isoxazole, 4-bromo-isoxazole, zlchem 1329 |
PubChem CID | 3862248 |
IUPAC Name | 4-bromo-1,2-oxazole |
SMILES | C1=C(C=NO1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15579573
|
Thermo Scientific Acros
458730010 |
1 g |
29.45 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15589573
|
Thermo Scientific Acros
458730050 |
5 g |
91.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97925-43-4 | |
147.96 | |
4-bromoisoxazole, 4-bormoisoxazole, isoxazole, 4-bromo, isoxazole, 4-bromo-6ci,9ci, 4-bromo isoxazole, 4-bromo-isoxazole, zlchem 1329 | |
4-bromo-1,2-oxazole |
C3H2BrNO | |
IRDVIGFQMLUJAM-UHFFFAOYSA-N | |
3862248 | |
C1=C(C=NO1)Br |
Specifications
97925-43-4 | |
Authentic | |
Glass Bottle | |
4-bromoisoxazole, 4-bormoisoxazole, isoxazole, 4-bromo, isoxazole, 4-bromo-6ci,9ci, 4-bromo isoxazole, 4-bromo-isoxazole, zlchem 1329 | |
C1=C(C=NO1)Br | |
147.96 | |
147.96 | |
4-Bromoisoxazole |
39.0°C to 41.0°C | |
94% min. (GC) | |
C3H2BrNO | |
IRDVIGFQMLUJAM-UHFFFAOYSA-N | |
4-bromo-1,2-oxazole | |
3862248 | |
95% |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
GHS P Statement Wear protective gloves/protective clothing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
- 4-Bromoisoxazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.