missing translation for 'onlineSavingsMsg'
Learn More

4-Bromofuran-2-carboxamide, 96%, Thermo Scientific Chemicals

Product Code. 15429027
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of 6-Phenanthridinones. Bromofuran-2-carboxamide used in the investigation of supramolecular amide and thioamide synthons in hydrogen bonding patterns.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957345-95-8
Molecular Formula C5H4BrNO2
Molecular Weight (g/mol) 189.996
MDL Number MFCD12755858
InChI Key UDNSRLLRTQICFU-UHFFFAOYSA-N
Synonym 4-bromo-furan-2-carboxylic acid amide
PubChem CID 57589975
IUPAC Name 4-bromofuran-2-carboxamide
SMILES C1=C(OC=C1Br)C(=O)N

Specifications

Quantity 1 g
Formula Weight 189.99
Percent Purity 96%
Chemical Name or Material 4-Bromofuran-2-carboxamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'