missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobutyl acetate, 97%, Thermo Scientific Chemicals
46.03 € - 559.00 €
Chemical Identifiers
CAS | 4753-59-7 |
---|---|
Molecular Formula | C6H11BrO2 |
Molecular Weight (g/mol) | 195.056 |
MDL Number | MFCD00000263 |
InChI Key | UOABIRUEGSGTSA-UHFFFAOYSA-N |
Synonym | 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide |
PubChem CID | 78491 |
IUPAC Name | 4-bromobutyl acetate |
SMILES | CC(=O)OCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447527
|
Thermo Scientific Alfa Aesar
A15632.09 |
10 g |
46.03 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457527
|
Thermo Scientific Alfa Aesar
A15632.18 |
50 g |
150.75 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467527
|
Thermo Scientific Alfa Aesar
A15632.30 |
250 g |
559.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4753-59-7 | |
195.056 | |
UOABIRUEGSGTSA-UHFFFAOYSA-N | |
78491 | |
CC(=O)OCCCCBr |
C6H11BrO2 | |
MFCD00000263 | |
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
4-bromobutyl acetate |
Specifications
4753-59-7 | |
96°C to 97°C (15 mmHg) | |
C6H11BrO2 | |
MFCD00000263 | |
4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide | |
CC(=O)OCCCCBr | |
195.056 | |
195.07 | |
4-Bromobutyl acetate |
1.348 | |
109°C (228°F) | |
1.46 | |
1749695 | |
UOABIRUEGSGTSA-UHFFFAOYSA-N | |
4-bromobutyl acetate | |
78491 | |
≥98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 225-277-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only