missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobutoxy)benzene, 97%, Thermo Scientific Chemicals

Product Code. 11437157
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1200-03-9
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00000262
InChI Key QBLISOIWPZSVIK-UHFFFAOYSA-N
Synonym 4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol
PubChem CID 70986
IUPAC Name 4-bromobutoxybenzene
SMILES BrCCCCOC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 153°C to 154°C (18 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 25 g
Beilstein 2044374
Solubility Information Insoluble in water.
Formula Weight 229.12
Percent Purity 97%
Chemical Name or Material (4-Bromobutoxy)benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.