Learn More
4-bromo-2-fluorobenzyl Chloride, 98%, Thermo Scientific™
37.50 € - 110.00 €
Chemical Identifiers
CAS | 85510-82-3 |
---|---|
Molecular Formula | C7H5BrClF |
Molecular Weight (g/mol) | 223.47 |
MDL Number | MFCD02094036 |
InChI Key | UDKQGFMDBMYVHI-UHFFFAOYSA-N |
Synonym | 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene |
PubChem CID | 2773361 |
IUPAC Name | 4-bromo-1-(chloromethyl)-2-fluorobenzene |
SMILES | FC1=CC(Br)=CC=C1CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10230493
|
Thermo Scientific Acros
384460050 |
5 g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10553143
|
Thermo Scientific Acros
384460250 |
25 g | Glass bottle |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Chemical Identifiers
85510-82-3 | |
223.47 | |
UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
2773361 | |
FC1=CC(Br)=CC=C1CCl |
C7H5BrClF | |
MFCD02094036 | |
4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
4-bromo-1-(chloromethyl)-2-fluorobenzene |
Specifications
85510-82-3 | |
100.0 | |
Authentic | |
C7H5BrClF | |
1.5610 to 1.5630 (20°C, 589nm) | |
4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene | |
FC1=CC(Br)=CC=C1CCl | |
223.47 | |
223.47 | |
Liquid |
97.5 | |
Colorless | |
97.5% min. (GC) | |
ClCH2C6H3(F)Br | |
MFCD02094036 | |
UDKQGFMDBMYVHI-UHFFFAOYSA-N | |
4-bromo-1-(chloromethyl)-2-fluorobenzene | |
2773361 | |
98% | |
4-Bromo-2-fluorobenzyl chloride |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- 4-Bromo-2-fluorobenzyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only