missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dinitrotoluene, 97%, Thermo Scientific Chemicals

Product Code. 11426590
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95192-64-6
Molecular Formula C7H5BrN2O4
Molecular Weight (g/mol) 261.031
MDL Number MFCD07357280
InChI Key UOGCLPDKGPPDHM-UHFFFAOYSA-N
Synonym 4-bromo-2,6-dinitrotoluene, 5-bromo-1,3-dinitro-2-methylbenzene, 5-bromo-2-methyl-1,3-dinitro-benzene, benzene, 5-bromo-2-methyl-1,3-dinitro, 2,6-dinitro-4-bromo-tolurene, 5-bromanyl-2-methyl-1,3-dinitro-benzene
PubChem CID 13443186
IUPAC Name 5-bromo-2-methyl-1,3-dinitrobenzene
SMILES CC1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]

Specifications

Melting Point 86°C to 89°C
Quantity 1 g
UN Number UN2811
Formula Weight 261.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,6-dinitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'