missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2,6-difluorobenzonitrile, 95%

Product Code. 15435557
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 123843-67-4
Molecular Formula C7H2BrF2N
Molecular Weight (g/mol) 218.00
MDL Number MFCD01310981
InChI Key TZHQWUAOIWRFSW-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluoro-benzonitrile, 2,6-difluoro-4-bromobenzonitrile, benzonitrile, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluorobenzenecarbonitrile, pubchem4699, acmc-1bymk, ksc513m5d, 4-bromo-2,6-difluoro-bezonitrile, 2,6-difluoro-4-bromo-benzonitrile
PubChem CID 2756916
IUPAC Name 4-bromo-2,6-difluorobenzonitrile
SMILES FC1=CC(Br)=CC(F)=C1C#N

Specifications

Melting Point 79°C to 82°C
Quantity 1 g
UN Number UN3439
Formula Weight 218
Percent Purity 95%
Chemical Name or Material 4-Bromo-2,6-difluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'