Learn More
4-Bromo-2,5-difluorophenol, 99%, Thermo Scientific Chemicals
82.80 € - 266.00 €
Chemical Identifiers
CAS | 486424-36-6 |
---|---|
Molecular Formula | C6H3BrF2O |
Molecular Weight (g/mol) | 208.99 |
MDL Number | MFCD04973751 |
InChI Key | BYZMZJIWCQTYSR-UHFFFAOYSA-N |
Synonym | 1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene |
PubChem CID | 7018042 |
IUPAC Name | 4-bromo-2,5-difluorophenol |
SMILES | C1=C(C(=CC(=C1F)Br)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346639
|
Thermo Scientific Alfa Aesar
H26212.06 |
5 g |
82.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356639
|
Thermo Scientific Alfa Aesar
H26212.14 |
25 g |
266.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.
Solubility
Soluble in water (slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
486424-36-6 | |
208.99 | |
BYZMZJIWCQTYSR-UHFFFAOYSA-N | |
7018042 | |
C1=C(C(=CC(=C1F)Br)F)O |
C6H3BrF2O | |
MFCD04973751 | |
1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene | |
4-bromo-2,5-difluorophenol |
Specifications
486424-36-6 | |
C6H3BrF2O | |
1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene | |
BYZMZJIWCQTYSR-UHFFFAOYSA-N | |
4-bromo-2,5-difluorophenol | |
7018042 | |
99% |
51°C to 53°C | |
MFCD04973751 | |
Soluble in water (slightly). | |
C1=C(C(=CC(=C1F)Br)F)O | |
208.99 | |
208.99 | |
4-Bromo-2,5-difluorophenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.