missing translation for 'onlineSavingsMsg'
Learn More

4-Bis(2-cyano-3-pyridylmethyl)aminobenzonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1421261-92-8
Molecular Formula C21H14N6
Molecular Weight (g/mol) 350.385
MDL Number MFCD22682981
InChI Key FPDANHUWWTXQDQ-UHFFFAOYSA-N
Synonym 3-4-cyanophenyl 2-cyanopyridin-3-yl methyl amino methyl pyridine-2-carbonitrile, 4-bis 2-cyano-3-pyridylmethyl aminobenzonitrile
PubChem CID 97030352
IUPAC Name 3-[[4-cyano-N-[(2-cyanopyridin-3-yl)methyl]anilino]methyl]pyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)CN(CC2=C(N=CC=C2)C#N)C3=CC=C(C=C3)C#N
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1421261-92-8
350.385
FPDANHUWWTXQDQ-UHFFFAOYSA-N
97030352
C1=CC(=C(N=C1)C#N)CN(CC2=C(N=CC=C2)C#N)C3=CC=C(C=C3)C#N
C21H14N6
MFCD22682981
3-4-cyanophenyl 2-cyanopyridin-3-yl methyl amino methyl pyridine-2-carbonitrile, 4-bis 2-cyano-3-pyridylmethyl aminobenzonitrile
3-[[4-cyano-N-[(2-cyanopyridin-3-yl)methyl]anilino]methyl]pyridine-2-carbonitrile

Specifications

1421261-92-8
MFCD22682981
3-4-cyanophenyl 2-cyanopyridin-3-yl methyl amino methyl pyridine-2-carbonitrile, 4-bis 2-cyano-3-pyridylmethyl aminobenzonitrile
C1=CC(=C(N=C1)C#N)CN(CC2=C(N=CC=C2)C#N)C3=CC=C(C=C3)C#N
350.385
350.38
4-Bis(2-cyano-3-pyridylmethyl)aminobenzonitrile
C21H14N6
UN3439
FPDANHUWWTXQDQ-UHFFFAOYSA-N
3-[[4-cyano-N-[(2-cyanopyridin-3-yl)methyl]anilino]methyl]pyridine-2-carbonitrile
97030352
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications