missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxypyridine N-oxide, 98%, Thermo Scientific Chemicals

14.27 € - 47.64 €

Chemical Identifiers

CAS 2683-66-1
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.23
MDL Number MFCD00047427
InChI Key SUSQPKJQYWTFPU-UHFFFAOYSA-N
Synonym 4-benzyloxy pyridine n-oxide, 4-benzyloxy pyridine 1-oxide, 4-benzyloxypyridine n-oxide, 4-benzyloxypyridine 1-oxide, 4-benzyloxypyridine-n-oxide, 4-benzyloxy pyridin-1-ium-1-olate, pyridine, 4-phenylmethoxy-, 1-oxide, 4-phenylmethoxy pyridin-1-ol, pubchem2123
PubChem CID 295871
SMILES [O-][N+]1=CC=C(OCC2=CC=CC=C2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334835
View Documents
Thermo Scientific Alfa Aesar
L17946.03
1 g
14.27 €
1g
Please to purchase this item. Need a web account? Register with us today!
11344835
View Documents
Thermo Scientific Alfa Aesar
L17946.06
5 g
47.64 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2683-66-1
201.23
SUSQPKJQYWTFPU-UHFFFAOYSA-N
295871
C12H11NO2
MFCD00047427
4-benzyloxy pyridine n-oxide, 4-benzyloxy pyridine 1-oxide, 4-benzyloxypyridine n-oxide, 4-benzyloxypyridine 1-oxide, 4-benzyloxypyridine-n-oxide, 4-benzyloxy pyridin-1-ium-1-olate, pyridine, 4-phenylmethoxy-, 1-oxide, 4-phenylmethoxy pyridin-1-ol, pubchem2123
[O-][N+]1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

2683-66-1
C12H11NO2
165005
SUSQPKJQYWTFPU-UHFFFAOYSA-N
4-(benzyloxy)pyridin-1-ium-1-olate
295871
98%
173°C to 175°C
MFCD00047427
4-benzyloxy pyridine n-oxide, 4-benzyloxy pyridine 1-oxide, 4-benzyloxypyridine n-oxide, 4-benzyloxypyridine 1-oxide, 4-benzyloxypyridine-n-oxide, 4-benzyloxy pyridin-1-ium-1-olate, pyridine, 4-phenylmethoxy-, 1-oxide, 4-phenylmethoxy pyridin-1-ol, pubchem2123
[O-][N+]1=CC=C(OCC2=CC=CC=C2)C=C1
201.23
201.22
4-Benzyloxypyridine N-oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.