missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Benzyloxycarbonylamino)cyclohexanone, 97%, Thermo Scientific™
234.00 €
Chemical Identifiers
CAS | 16801-63-1 |
---|---|
Molecular Formula | C14H17NO3 |
Molecular Weight (g/mol) | 247.29 |
MDL Number | MFCD06658393 |
InChI Key | VHXBIBFCJISKFA-UHFFFAOYSA-N |
Synonym | benzyl 4-oxocyclohexyl carbamate, 4-n-cbz-cyclohexanone, 4-n-cbz-amino-cyclohexanone, 4-oxocyclohexyl carbamic acid benzyl ester, 4-n-cbz-aminocyclohexanone, 4-benzyloxycarbonylamino cyclohexanone, 4-cbz-amino cyclohexanone, n-cbz-4-amino-cyclohexanone, benzyl n-4-oxocyclohexyl carbamate, n-cbz-4-aminocyclohexanone |
PubChem CID | 14615292 |
IUPAC Name | benzyl N-(4-oxocyclohexyl)carbamate |
SMILES | O=C(NC1CCC(=O)CC1)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11341929
|
Thermo Scientific Alfa Aesar
|
1 g |
N/A
|
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11351929
|
Thermo Scientific Alfa Aesar
H26015.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
16801-63-1 | |
247.29 | |
VHXBIBFCJISKFA-UHFFFAOYSA-N | |
14615292 | |
O=C(NC1CCC(=O)CC1)OCC1=CC=CC=C1 |
C14H17NO3 | |
MFCD06658393 | |
benzyl 4-oxocyclohexyl carbamate, 4-n-cbz-cyclohexanone, 4-n-cbz-amino-cyclohexanone, 4-oxocyclohexyl carbamic acid benzyl ester, 4-n-cbz-aminocyclohexanone, 4-benzyloxycarbonylamino cyclohexanone, 4-cbz-amino cyclohexanone, n-cbz-4-amino-cyclohexanone, benzyl n-4-oxocyclohexyl carbamate, n-cbz-4-aminocyclohexanone | |
benzyl N-(4-oxocyclohexyl)carbamate |
Specifications
16801-63-1 | |
Odorless | |
MFCD06658393 | |
VHXBIBFCJISKFA-UHFFFAOYSA-N | |
benzyl N-(4-oxocyclohexyl)carbamate | |
14615292 | |
97% |
81°C to 83°C | |
C14H17NO3 | |
benzyl 4-oxocyclohexyl carbamate, 4-n-cbz-cyclohexanone, 4-n-cbz-amino-cyclohexanone, 4-oxocyclohexyl carbamic acid benzyl ester, 4-n-cbz-aminocyclohexanone, 4-benzyloxycarbonylamino cyclohexanone, 4-cbz-amino cyclohexanone, n-cbz-4-amino-cyclohexanone, benzyl n-4-oxocyclohexyl carbamate, n-cbz-4-aminocyclohexanone | |
O=C(NC1CCC(=O)CC1)OCC1=CC=CC=C1 | |
247.29 | |
247.29 | |
4-(Benzyloxycarbonylamino)cyclohexanone |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title