Learn More
4-Benzyloxybenzeneboronic acid, 97%, Thermo Scientific™
21.91 € - 147.00 €
Chemical Identifiers
CAS | 146631-00-7 |
---|---|
Molecular Formula | C13H13BO3 |
Molecular Weight (g/mol) | 228.054 |
MDL Number | MFCD01075705 |
InChI Key | DMJHEIDWSIAXCS-UHFFFAOYSA-N |
Synonym | 4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561 |
PubChem CID | 2734314 |
IUPAC Name | (4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405787
|
Thermo Scientific Alfa Aesar
B24351.01 |
0.25 g |
21.91 €
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485777
|
Thermo Scientific Alfa Aesar
B24351.03 |
1 g | N/A | ||||||
11495777
|
Thermo Scientific Alfa Aesar
B24351.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as electronic intermediate. It is also utilized in HPLC and NMR spectroscopy.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
146631-00-7 | |
228.054 | |
DMJHEIDWSIAXCS-UHFFFAOYSA-N | |
2734314 | |
B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O |
C13H13BO3 | |
MFCD01075705 | |
4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561 | |
(4-phenylmethoxyphenyl)boronic acid |
Specifications
146631-00-7 | |
C13H13BO3 | |
4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561 | |
B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O | |
228.054 | |
228.06 | |
4-Benzyloxybenzeneboronic acid |
193°C to 198°C | |
MFCD01075705 | |
DMJHEIDWSIAXCS-UHFFFAOYSA-N | |
(4-phenylmethoxyphenyl)boronic acid | |
2734314 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures