missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomorpholine, 95%, Thermo Scientific Chemicals

Product Code. 11479327
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4319-49-7
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00006174
InChI Key MKQLBNJQQZRQJU-UHFFFAOYSA-N
Synonym 4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
PubChem CID 20315
IUPAC Name morpholin-4-amine
SMILES C1COCCN1N

Specifications

Density 1.059
Boiling Point 168°C
Flash Point 58°C (137°F)
Refractive Index 1.4785
Quantity 25 g
UN Number UN2733
Sensitivity Air sensitive
Formula Weight 102.14
Percent Purity 98%
Chemical Name or Material 4-Aminomorpholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.