missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoimidazole-5-carboxamide hydrochloride, 98%, Thermo Scientific Chemicals
22.20 € - 242.00 €
Chemical Identifiers
CAS | 72-40-2 |
---|---|
Molecular Formula | C4H7ClN4O |
Molecular Weight (g/mol) | 162.58 |
MDL Number | MFCD00012704 |
InChI Key | MXCUYSMIELHIQL-UHFFFAOYSA-N |
Synonym | 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride |
PubChem CID | 66146 |
IUPAC Name | 4-amino-1H-imidazole-5-carboxamide;hydrochloride |
SMILES | Cl.NC(=O)C1=C(N)N=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11416926
|
Thermo Scientific Alfa Aesar
B24012.03 |
1 g | N/A | ||||||
11426926
|
Thermo Scientific Alfa Aesar
B24012.06 |
5 g |
74.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436926
|
Thermo Scientific Alfa Aesar
B24012.14 |
25 g |
242.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72-40-2 | |
162.58 | |
MXCUYSMIELHIQL-UHFFFAOYSA-N | |
66146 | |
Cl.NC(=O)C1=C(N)N=CN1 |
C4H7ClN4O | |
MFCD00012704 | |
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride | |
4-amino-1H-imidazole-5-carboxamide;hydrochloride |
Specifications
72-40-2 | |
Odorless | |
MFCD00012704 | |
Hygroscopic | |
MXCUYSMIELHIQL-UHFFFAOYSA-N | |
4-amino-1H-imidazole-5-carboxamide;hydrochloride | |
66146 | |
98% |
∼270°C (decomposition) | |
C4H7ClN4O | |
3701645 | |
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride | |
Cl.NC(=O)C1=C(N)N=CN1 | |
162.58 | |
162.58 | |
4-Aminoimidazole-5-carboxamide hydrochloride |
Safety and Handling
missing translation for 'einecsNumber' : 200-778-3
missing translation for 'rtecsNumber' : NI3911000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only