missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15459745
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819058-34-9
Molecular Formula C12H17BFNO2
Molecular Weight (g/mol) 237.08
MDL Number MFCD09033884
InChI Key AIXGNRNTXUKZLC-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronic acid pinacol ester, 4-amino-3-fluorophenylboronic acid, pinacol ester, benzenamine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-amino-3-fluorophenylboronicacidpinacolester, 4-amino-3-fluorophenylboronic acid,pinacol ester, 4-amino-3-fluorobenzeneboronic acid pinacol ester, 4-amino-3-fluorophenyl boronic acid pinacol ester, 2-3-fluoro-4-aminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17906172
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 237.08
Percent Purity 96%
Chemical Name or Material 4-Amino-3-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'