Learn More
4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride, Thermo Scientific™
96.65 € - 311.55 €
Chemical Identifiers
CAS | 1301739-00-3 |
---|---|
Molecular Formula | C11H18ClN3O |
Molecular Weight (g/mol) | 243.735 |
MDL Number | MFCD11869912 |
InChI Key | QBVBNVDHWTXFAT-UHFFFAOYSA-N |
Synonym | 4-amino-1-3-methoxy-2-pyridyl piperidine hydrochloride, 1-3-methoxypyridin-2-yl piperidin-4-amine hydrochloride, 2-4-amino-1-piperidinyl-3-methoxypyridine, 1-3-methoxy-2-pyridyl-4-piperidinamine hydrochloride |
PubChem CID | 53400829 |
IUPAC Name | 1-(3-methoxypyridin-2-yl)piperidin-4-amine;hydrochloride |
SMILES | COC1=C(N=CC=C1)N2CCC(CC2)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479205
|
Thermo Scientific Alfa Aesar
H51176.MD |
250 mg | N/A | ||||||
15469205
|
Thermo Scientific Alfa Aesar
H51176.03 |
1 g |
311.55 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1301739-00-3 | |
243.735 | |
QBVBNVDHWTXFAT-UHFFFAOYSA-N | |
53400829 | |
COC1=C(N=CC=C1)N2CCC(CC2)N.Cl |
C11H18ClN3O | |
MFCD11869912 | |
4-amino-1-3-methoxy-2-pyridyl piperidine hydrochloride, 1-3-methoxypyridin-2-yl piperidin-4-amine hydrochloride, 2-4-amino-1-piperidinyl-3-methoxypyridine, 1-3-methoxy-2-pyridyl-4-piperidinamine hydrochloride | |
1-(3-methoxypyridin-2-yl)piperidin-4-amine;hydrochloride |
Specifications
1301739-00-3 | |
MFCD11869912 | |
QBVBNVDHWTXFAT-UHFFFAOYSA-N | |
1-(3-methoxypyridin-2-yl)piperidin-4-amine;hydrochloride | |
53400829 | |
4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride |
C11H18ClN3O | |
4-amino-1-3-methoxy-2-pyridyl piperidine hydrochloride, 1-3-methoxypyridin-2-yl piperidin-4-amine hydrochloride, 2-4-amino-1-piperidinyl-3-methoxypyridine, 1-3-methoxy-2-pyridyl-4-piperidinamine hydrochloride | |
COC1=C(N=CC=C1)N2CCC(CC2)N.Cl | |
243.735 | |
243.73 |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures