missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenoxyacetic acid, 98+%, Thermo Scientific™

Product Code. 11350034
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-81-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014363
InChI Key KMXZEXUYXUMHEQ-UHFFFAOYSA-N
Synonym 4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy
PubChem CID 74655
IUPAC Name 2-(4-acetylphenoxy)acetic acid
SMILES CC(=O)C1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 172°C to 174°C
Quantity 5 g
Beilstein 1956136
Formula Weight 194.19
Percent Purity ≥98%
Chemical Name or Material 4-Acetylphenoxyacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.