missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylbiphenyl, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 117°C to 123°C |
Boiling Point | 168°C (8 mmHg) |
Quantity | 100 g |
Beilstein | 1101615 |
Solubility Information | Insoluble in water. |
Formula Weight | 196.25 |
Percent Purity | 98% |
Chemical Name or Material | 4-Acetylbiphenyl |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction