missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Description
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is used as a phase transfer catalysts by transferring ions from one phase to another. It is involved in the synthesis of alkalides and electrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Yellow |
Density | 1.11 g/mL |
Refractive Index | 1.501 |
Quantity | 2 g |
Beilstein | 616444 |
Sensitivity | Light sensitive |
Solubility Information | Miscible with water. |
Formula Weight | 332.44 |
Percent Purity | 97% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'