missing translation for 'onlineSavingsMsg'
Learn More

4,5-Difluoro-2-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 15402198
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78056-39-0
Molecular Formula C6H4F2N2O2
Molecular Weight (g/mol) 174.11
MDL Number MFCD00010876
InChI Key WDMCABATCGQAKK-UHFFFAOYSA-N
Synonym 3,4-difluoro-6-nitroaniline, benzenamine, 4,5-difluoro-2-nitro, 4,5-difluoro-2-nitro-aniline, 4,5-difluoro-2-nitrophenylamine, 2-nitro-4,5-difluoroaniline, 2-amino-4,5-difluoronitrobenzene, 4,5-difluoro-2-nitro-phenylamine, 4,5-difluoro-2-nitrobenzenamine, pubchem2121, acmc-209pdb
PubChem CID 157151
IUPAC Name 4,5-difluoro-2-nitroaniline
SMILES NC1=CC(F)=C(F)C=C1[N+]([O-])=O

Specifications

Melting Point 107°C to 108°C
Quantity 5 g
Beilstein 2723242
Formula Weight 174.11
Percent Purity 98%
Chemical Name or Material 4,5-Difluoro-2-nitroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'