missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyanoimidazole, 99%, Thermo Scientific Chemicals

39.09 € - 119.60 €

Chemical Identifiers

CAS 1122-28-7
Molecular Formula C5H2N4
Molecular Weight (g/mol) 118.10
MDL Number MFCD00005194
InChI Key XGDRLCRGKUCBQL-UHFFFAOYSA-N
PubChem CID 70729
IUPAC Name 1H-imidazole-4,5-dicarbonitrile
SMILES N#CC1=C(N=CN1)C#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10455682
View Documents Promotion Details
Thermo Scientific Acros
368530050
5 g Glass bottle
39.09 €
5g
Please to purchase this item. Need a web account? Register with us today!
10212342
View Documents Promotion Details
Thermo Scientific Acros
368530250
25 g Glass bottle
119.60 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1122-28-7
118.10
XGDRLCRGKUCBQL-UHFFFAOYSA-N
1H-imidazole-4,5-dicarbonitrile
C5H2N4
MFCD00005194
70729
N#CC1=C(N=CN1)C#N

Specifications

1122-28-7
100.0
White
98.5% min. (HPLC)
MFCD00005194
XGDRLCRGKUCBQL-UHFFFAOYSA-N
1H-imidazole-4,5-dicarbonitrile
70729
99%
4, 5-Dicyanoimidazole, 99%
98.5
174.0°C to 177.0°C
Authentic
C5H2N4
4,5-dicyanoimidazole, imidazole-4,5-dicarbonitrile, 4,5-imidazoledicarbonitrile, activator, pubchem7602, 4, 5-dicyanoimidazole, moa 7, imidazole-4,5-dinitrile, acmc-1brr7, 4,5-dicyano-1h-imidazole
N#CC1=C(N=CN1)C#N
118.10
118.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Immediately call a POISON CENTER or

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 214-344-6

Product Identifier
  • 4,5-Dicyanoimidazole

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only