Learn More
4,5-Dichloroimidazole, 98%, Thermo Scientific Chemicals
46.10 € - 173.00 €
Chemical Identifiers
CAS | 15965-30-7 |
---|---|
Molecular Formula | C3H2Cl2N2 |
Molecular Weight (g/mol) | 136.96 |
MDL Number | MFCD00005195 |
InChI Key | QAJJXHRQPLATMK-UHFFFAOYSA-N |
Synonym | 4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa |
PubChem CID | 140018 |
IUPAC Name | 4,5-dichloro-1H-imidazole |
SMILES | ClC1=C(Cl)N=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362436
|
Thermo Scientific Alfa Aesar
L06262.06 |
5 g |
46.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372436
|
Thermo Scientific Alfa Aesar
L06262.14 |
25 g |
173.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Dichloroimidazole has been used in the synthesis of: phosphinite complexes, 1-hexyl-3-methyl-4,5-dichloroimidazolium iodide, precursor required for the preparation of silver-carbene complex-loaded into L-tyrosine polyphosphate nanoparticles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, strong acids.
Chemical Identifiers
15965-30-7 | |
136.96 | |
QAJJXHRQPLATMK-UHFFFAOYSA-N | |
140018 | |
ClC1=C(Cl)N=CN1 |
C3H2Cl2N2 | |
MFCD00005195 | |
4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa | |
4,5-dichloro-1H-imidazole |
Specifications
15965-30-7 | |
C3H2Cl2N2 | |
606155 | |
Insoluble in water. | |
ClC1=C(Cl)N=CN1 | |
136.96 | |
136.97 | |
4,5-Dichloroimidazole |
179°C to 182°C | |
MFCD00005195 | |
4,5-dichloroimidazole, 1h-imidazole, 4,5-dichloro, 4,5-dichlorimidazole, pubchem12880, acmc-1bqkp, 3,4-dichloro-2h-pyrazole, ksc530c6j, 1h-imidazole,4,5-dichloro, qajjxhrqplatmk-uhfffaoysa | |
QAJJXHRQPLATMK-UHFFFAOYSA-N | |
4,5-dichloro-1H-imidazole | |
140018 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only