missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylpiperazino)benzoic acid, 97%, Thermo Scientific™

Product Code. 10079101
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 86620-62-4
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD02682063
InChI Key UCFZVQHKTRSZMM-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl benzoic acid, 4-4-methylpiperazino benzoic acid, 4-4-methyl-piperazin-1-yl-benzoic acid, 4-4-methylpiperazinyl benzoic acid, 4-4-methyl-piperazino benzoic acid, benzoic acid, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzoic acid, pubchem10481, ksc448o3p
PubChem CID 736532
IUPAC Name 4-(4-methylpiperazin-1-yl)benzoic acid
SMILES CN1CCN(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazino)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.