Learn More
4,4-Dimethylcyclohexanone, 97%, Thermo Scientific Chemicals
25.42 € - 261.00 €
Chemical Identifiers
CAS | 4255-62-3 |
---|---|
InChI Key | PXQMSTLNSHMSJB-UHFFFAOYSA-N |
Synonym | 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanoneShow More |
PubChem CID | 138166 |
IUPAC Name | 4,4-dimethylcyclohexan-1-one |
SMILES | CC1(CCC(=O)CC1)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
13488628
|
Thermo Scientific Acros
448812500 |
250 mg | Glass bottle | N/A | N/A | N/A | |||||
13468628
|
Thermo Scientific Acros
448810010 |
1 g | Glass bottle |
40.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
13478628
|
Thermo Scientific Acros
448810050 |
5 g | Glass Bottle | N/A | N/A | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4255-62-3 | |
4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanoneShow More | |
4,4-dimethylcyclohexan-1-one |
PXQMSTLNSHMSJB-UHFFFAOYSA-N | |
138166 | |
CC1(CCC(=O)CC1)C |
Specifications
4255-62-3 | |
73°C (14.0 mmHg) | |
96% min. (GC) | |
(CH3)2C6H8(=O) | |
Solubility in water: insoluble | |
CC1(CCC(=O)CC1)C | |
126.2 | |
126.2 | |
4, 4-Dimethylcyclohexanone |
41°C to 43°C | |
62°C | |
C8H14O | |
4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone | |
PXQMSTLNSHMSJB-UHFFFAOYSA-N | |
4,4-dimethylcyclohexan-1-one | |
138166 | |
97% |
Safety and Handling
GHS Signal Word: Danger
- 4,4-Dimethylcyclohexanone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
Hazard Statement
- H319-Causes serious eye irritation.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.