missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenoxy)piperidine,98%, Thermo Scientific Chemicals

Product Code. 11409690
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97839-99-1
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.69
MDL Number MFCD04113544
InChI Key VOQMPZXAFLPTMM-UHFFFAOYSA-N
PubChem CID 5199889
IUPAC Name 4-(4-chlorophenoxy)piperidine
SMILES ClC1=CC=C(OC2CCNCC2)C=C1

Specifications

Quantity 1 g
Synonym 4-4-chlorophenoxy piperidine, 4-4-chloro-phenoxy-piperidine, piperidine, 4-4-chlorophenoxy, acmc-20amwb, 4-p-chlorophenoxy piperidine, 4-4-chlorophenyl oxy piperidine, 4-chloro-1-4-piperidyloxy benzene, piperidine,4-4-chlorophenoxy, piperidine, 4-4-chlorophenoxy-, hydrochloride
Solubility Information Slightly soluble in water.
Formula Weight 211.69
Percent Purity 98%
Chemical Name or Material 4-(4-Chlorophenoxy)piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'